missing translation for 'onlineSavingsMsg'
Learn More
Potassium carbonate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals Product Code.: 15139293

Potassium carbonate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals

Product Code. 15139293
10 kg, Plastic Drum
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 15139293

Brand: Thermo Scientific Acros 424080100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891.0°C
Color White
Physical Form Fine Granular Powder
Quantity 10 kg
Assay Percent Range 99+%
Linear Formula K2CO3
Fieser 01,1014; 04,50; 05,552; 08,408; 09,382; 10,323; 11,433; 12,403; 13,233; 15,167
Merck Index 15, 7740
Solubility Information Solubility in water: 1120g/L (20°C). Other solubilities: practically insoluble in ethanol, insoluble in acetone
IUPAC Name dipotassium carbonate
Formula Weight 138.21
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic Drum
Chemical Name or Material Potassium Carbonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468628 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog Number424085000,424080100,424081000,424080025
Lot NumberA0464860 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog Number424085000,424080100,424081000,424080025
Lot NumberB0155882 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number424085000,424080100,424081000,424080025
Lot NumberA0462316 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number424085000,424080100,424081000,424080025
Lot NumberA0460789 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number424085000,424080100,424081000,424080025
compliance-icons
Product Identifier
  • Potassium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium carbonate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.