missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, ACS, 99.0% min

Product Code. 11337457
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
25g
500g
5kg
This item is not returnable. View return policy

Product Code. 11337457

missing translation for 'mfr': Thermo Scientific Alfa Aesar 012609.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 584-08-7 | CK2O3 | 138.21 g/mol

It may be used in pharmaceutical laboratories as a drying agent or as a source of potassium. It is also used in fire extinguishers, to make soap, glass and to soften water. Aqueous potassium carbonate plays an important role in carbon dioxide absorption promoted by piperazine. It is also essential for the absorption of carbon dioxide promoted by boric acid. It is also useful for the synthesis of azoles and diazines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Physical Form Granules
Quantity 1 kg
Beilstein 4267587
Merck Index 14,7619
Solubility Information Soluble in water. Insoluble in ethanol.
IUPAC Name dipotassium;carbonate
Formula Weight 138.21
Percent Purity ≥99.0%
Grade ACS
Sensitivity Hygroscopic
Density 2.29
Chemical Name or Material Potassium carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.