missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™
CK2O3, CAS Number-584-08-7, carbonate of potash, carbonic acid, dipotassium salt, dipotassium carbonate, kaliumcarbonat, pearl ash, potash, potassiumcarbonate, potassium carbonate, potassium carbonate, anhydrous, salt of tartar, 500g, White, 138.21g/mol, BWHMMNNQKKPAPP-UHFFFAOYSA-L
£52.30 - £351.00
Chemical Identifiers
CAS | 584-08-7 |
---|---|
Molecular Formula | CK2O3 |
Molecular Weight (g/mol) | 138.21 |
MDL Number | MFCD00011382 |
InChI Key | BWHMMNNQKKPAPP-UHFFFAOYSA-L |
Synonym | potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonateShow More |
PubChem CID | 11430 |
SMILES | [K+].[K+].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10647612
|
Fisher Chemical
P/4120/50 |
250 g |
£52.30
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10637422
|
Fisher Chemical
P/4120/53 |
500 g |
£83.95
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10191480
|
Fisher Chemical
P/4120/62 |
2.5 kg |
£351.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CK2O3, CAS Number-584-08-7, carbonate of potash, carbonic acid, dipotassium salt, dipotassium carbonate, kaliumcarbonat, pearl ash, potash, potassiumcarbonate, potassium carbonate, potassium carbonate, anhydrous, salt of tartar, 500g, White, 138.21g/mol, BWHMMNNQKKPAPP-UHFFFAOYSA-L
Chemical Identifiers
584-08-7 | |
138.21 | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
11430 |
CK2O3 | |
MFCD00011382 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonateShow More | |
[K+].[K+].[O-]C([O-])=O |
Specifications
891°C | |
White | |
Powder/Solid | |
MFCD00011382 | |
BWHMMNNQKKPAPP-UHFFFAOYSA-L | |
dipotassium;carbonate | |
11430 | |
Plastic powder jar |
584-08-7 | |
11.5 to 12 | |
CK2O3 | |
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate | |
[K+].[K+].[O-]C([O-])=O | |
138.21 | |
138.21g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium carbonate
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™