missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical

Product Code. 10497260 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10497260

Brand: Fisher Chemical P/4120/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Specifications

Melting Point 891°C
CAS 584-08-7
Color White
pH 11.5 to 12
Physical Form Powder/Solid
Quantity 1 kg
Molecular Formula CK2O3
MDL Number MFCD00011382
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES [K+].[K+].[O-]C([O-])=O
IUPAC Name dipotassium;carbonate
Molecular Weight (g/mol) 138.21
PubChem CID 11430
Formula Weight 138.21g/mol
Packaging Plastic powder jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.