missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, puriss p.a, ACS, ≥99.0% (T), Honeywell™ Fluka™

Catalog Number 15685910
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
25 kg
4 x 2.5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
584-08-7
CK2O3
138.21
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
11430
dipotassium;carbonate
[K+].[K+].[O-]C([O-])=O
This item is not returnable. View return policy
584-08-7
CK2O3
138.21
MFCD00011382
BWHMMNNQKKPAPP-UHFFFAOYSA-L
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
11430
dipotassium;carbonate
[K+].[K+].[O-]C([O-])=O

Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 891°C
Color Colorless or White
Quantity 2.5 kg
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
Formula Weight 138.21g/mol
Percent Purity ≥99.0% (T)
Packaging Poly bottle
Density 2.430g/cm3
Chemical Name or Material Potassium carbonate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'