missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, puriss p.a, ACS, ≥99.0% (T), Honeywell™ Fluka™

Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)

£28.01 - £1200.77

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
IUPAC Name dipotassium;carbonate
SMILES [K+].[K+].[O-]C([O-])=O
View More Specs

Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15695910
View Documents
Honeywell-Fluka
60109-250G
250 g Poly bottle
£28.01
250g
Please to purchase this item. Need a web account? Register with us today!
15625920
View Documents
Honeywell-Fluka
60109-500G
500 g Poly bottle
£51.78
500g
Please to purchase this item. Need a web account? Register with us today!
15675910
View Documents
Honeywell-Fluka
60109-1KG
1 kg Poly bottle
£96.00
1kg
Please to purchase this item. Need a web account? Register with us today!
15685910
View Documents
Honeywell-Fluka
60109-2.5KG
2.5 kg Poly bottle
£171.22
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15615920
View Documents
Honeywell-Fluka
60109-4X2.5KG
4 x 2.5 kg Poly bottle
£518.30
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15605920
View Documents
Honeywell Fluka
60109-25KG
25 kg Poly bottle
£1200.77
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

584-08-7
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430
[K+].[K+].[O-]C([O-])=O
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
dipotassium;carbonate

Specifications

891°C
Colorless or White
K2CO3
NONH for all modes of transport
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
[K+].[K+].[O-]C([O-])=O
138.21
138.21g/mol
2.430g/cm3
584-08-7
CK2O3
MFCD00011382
4267587
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium;carbonate
11430
≥99.0% (T)
Potassium carbonate
Videos
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

missing translation for 'rtecsNumber' : 209-529-3

Product Identifier
  • Potassium carbonate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Potassium carbonate

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.