missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, purum p.a., Honeywell™

Purum p.a., anhydrous,≥99.0% (T)

£55.02 - £169.88

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
IUPAC Name dipotassium;carbonate
SMILES [K+].[K+].[O-]C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15635920
View Documents
Honeywell Research Chemicals
60110-1KG
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15645920
View Documents
Honeywell Research Chemicals
60110-5KG
5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications

Chemical Identifiers

584-08-7
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430
[K+].[K+].[O-]C([O-])=O
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
dipotassium;carbonate

Specifications

584-08-7
K2CO3
NONH for all modes of transport
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
[K+].[K+].[O-]C([O-])=O
138.21
138.21g/mol
purum p.a.
CK2O3
MFCD00011382
4267587
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium;carbonate
11430
≥99% (T)
Potassium carbonate
Videos
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

missing translation for 'rtecsNumber' : TS7750000

SDS
Documents

Documents

Product Certifications