missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium carbonate, Reagent grade, Honeywell
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
12 kg
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 2.5 kg |
Linear Formula | K2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4267587 |
IUPAC Name | dipotassium carbonate |
Formula Weight | 138.21g/mol |
Percent Purity | ≥98% |
Grade | Reagent |
Chemical Name or Material | Potassium carbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'