missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, Reagent grade, Honeywell

Product Code. 15614410
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
12 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥98%, powder, −325 mesh

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Quantity 250 g
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
IUPAC Name dipotassium;carbonate
Formula Weight 138.21g/mol
Percent Purity ≥98%
Grade Reagent
Chemical Name or Material Potassium carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.