missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, ReagentPlus™, Honeywell™

Reagent Grade, 99%

£28.39 - £333.48

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15673160
View Documents
Honeywell Research Chemicals
310263-1KG
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15683160
View Documents
Honeywell Research Chemicals
310263-2.5KG
2.5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15693160
View Documents
Honeywell Research Chemicals
310263-5KG
5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15663160
View Documents
Honeywell Research Chemicals
310263-12KG
12 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

584-08-7
138.21
BWHMMNNQKKPAPP-UHFFFAOYSA-L
11430
CK2O3
MFCD00011382
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
[K+].[K+].[O-]C([O-])=O

Specifications

584-08-7
K2CO3
NONH for all modes of transport
potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
[K+].[K+].[O-]C([O-])=O
138.21
138.21g/mol
ReagentPlus™
CK2O3
MFCD00011382
4267587
BWHMMNNQKKPAPP-UHFFFAOYSA-L
dipotassium carbonate
11430
99%
Potassium Carbonate
Videos
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

H302-H315-H319-H335

missing translation for 'einecsNumber' : 209-529-3

missing translation for 'rtecsNumber' : TS7750000

SDS
Documents

Documents

Product Certifications