missing translation for 'onlineSavingsMsg'
Learn More

Potassium chlorate, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10076010
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
MDL Number MFCD00011361
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
PubChem CID 6426889
IUPAC Name potassium;chlorate
SMILES [O-]Cl(=O)=O.[K+]

Specifications

Melting Point 356.0°C to 368.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99+%
Linear Formula KClO3
Fieser 01,927
Merck Index 15, 7741
Solubility Information Solubility in water: 73g/L (20°C). Other solubilities: 555g/L hot water (100°C), soluble in alkalies, 1g/50mL glycerol, practically insoluble in alcohol, insoluble in ether
Formula Weight 122.55
Percent Purity ≥99%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Potassium chlorate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium chlorate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Oxidising solid Category 1
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'