Learn More
Chemical Identifiers
CAS | 3811-04-9 |
---|---|
Molecular Formula | ClKO3 |
Molecular Weight (g/mol) | 122.55 |
MDL Number | MFCD00011361 |
InChI Key | VKJKEPKFPUWCAS-UHFFFAOYSA-M |
Synonym | potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt |
PubChem CID | 6426889 |
IUPAC Name | potassium;chlorate |
SMILES | [O-]Cl(=O)=O.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11488936
|
Thermo Scientific Alfa Aesar
A17075.30 |
250 g |
£27.44
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478936
|
Thermo Scientific Alfa Aesar
A17075.0B |
1000 g |
£68.04
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11498936
|
Thermo Scientific Alfa Aesar
A17075.0I |
5000 g |
£213.06
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium chlorate is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium chlorate is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.
Solubility
Soluble in water, glycerol. Insoluble in alcohol.
Notes
Store away from incompatible materials. Store in a well-ventilated place.
Chemical Identifiers
3811-04-9 | |
122.55 | |
VKJKEPKFPUWCAS-UHFFFAOYSA-M | |
6426889 | |
[O-]Cl(=O)=O.[K+] |
ClKO3 | |
MFCD00011361 | |
potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt | |
potassium;chlorate |
Specifications
400°C (decomposition) | |
3811-04-9 | |
MFCD00011361 | |
14,7620 | |
Soluble in water,glycerol. Insoluble in alcohol. | |
[O-]Cl(=O)=O.[K+] | |
122.55 | |
122.55 | |
Odorless | |
2.32 |
368°C | |
ClKO3 | |
UN1485 | |
potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt | |
VKJKEPKFPUWCAS-UHFFFAOYSA-M | |
potassium;chlorate | |
6426889 | |
≥99% | |
Hygroscopic | |
Potassium chlorate |
Safety and Handling
GHS H Statement
H271-H302-H332
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
Harmful if inhaled.
P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
H271-H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM CHLORATE
missing translation for 'einecsNumber' : 223-289-7
missing translation for 'rtecsNumber' : FO0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Potassium chlorate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Oxidising solid Category 1
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
- MIXTURE LIST-Contains : Potassium chlorate
RUO – Research Use Only