Learn More
Potassium chlorate, ACS, 99.0% min, Thermo Scientific Chemicals
£65.22 - £234.17
Chemical Identifiers
CAS | 3811-04-9 |
---|---|
Molecular Formula | ClKO3 |
Molecular Weight (g/mol) | 122.55 |
MDL Number | MFCD00011361 |
InChI Key | VKJKEPKFPUWCAS-UHFFFAOYSA-M |
Synonym | potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt |
PubChem CID | 6426889 |
IUPAC Name | potassium;chlorate |
SMILES | [O-]Cl(=O)=O.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317958
|
Thermo Scientific Alfa Aesar
036494.36 |
500 g |
£65.22
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327958
|
Thermo Scientific Alfa Aesar
036494.A3 |
2 kg |
£234.17
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3811-04-9 | |
122.55 | |
VKJKEPKFPUWCAS-UHFFFAOYSA-M | |
6426889 | |
[O-]Cl(=O)=O.[K+] |
ClKO3 | |
MFCD00011361 | |
potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt | |
potassium;chlorate |
Specifications
400°C (decomposition) | |
3811-04-9 | |
99.0% | |
MFCD00011361 | |
Soluble in water,glycerol. Insoluble in alcohol. | |
[O-]Cl(=O)=O.[K+] | |
122.55 | |
122.55 | |
Hygroscopic | |
Potassium chlorate, For analysis ACS |
368°C | |
Crystalline | |
ClKO3 | |
potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt | |
VKJKEPKFPUWCAS-UHFFFAOYSA-M | |
potassium;chlorate | |
6426889 | |
ACS Reagent | |
2.32 g/mL |
Safety and Handling
P210-P220-P221-P261-P264b-P270-P271-P280-P283-P301+P312-P304+P340-P306+P360-P312-P330-P370+P378q-P371+P380+P375-P501c
H271-H302+H332
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 223-289-7
missing translation for 'rtecsNumber' : FO0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only