missing translation for 'onlineSavingsMsg'
Learn More
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals Product Code.: 11325417

Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals

Product Code. 11325417
4 L
Click to view available options
Quantity:
500 mL
1 L
4 L
This item is not returnable. View return policy

Product Code. 11325417

Brand: Thermo Scientific Alfa Aesar 042679.K7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium chloride 0.010M solution is used as conductive standard and as electrolyte in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7, 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Quantity 4 L
Odor Odorless
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Assay Percent Range 0.010M
Physical Form Liquid
Color Colorless
IUPAC Name potassium;chloride
Solubility Miscible with water
Formula Weight 74.55
Concentration or Composition (by Analyte or Components) Potassium chloride: 0.075%; Water: 99.925%
Chemical Name or Material Potassium chloride, 1413μmho at 25°C
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium chloride, 0.010M, Conductivity Standard, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.