missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, 1M aq. soln., RNAse free, Thermo Scientific Chemicals
Click to view available options
Quantity:
125 mL
250 mL
Description
It may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins. This solution is used in a variety of biological applications, including DNA and RNA isolation techniques. Also used to establish a constant ionic strength in a sample solution. pH: 5.0-8.0 (25°C, 1 M in H2O)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins. This solution is used in a variety of biological applications, including DNA and RNA isolation techniques. Also used to establish a constant ionic strength in a sample solution. pH: 5.0-8.0 (25°C, 1 M in H2O)
Solubility
Not miscible in water.
Notes
Store at room temperature. Ensure good ventilation.
It may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins. This solution is used in a variety of biological applications, including DNA and RNA isolation techniques. Also used to establish a constant ionic strength in a sample solution. pH: 5.0-8.0 (25°C, 1 M in H2O)
Solubility
Not miscible in water.
Notes
Store at room temperature. Ensure good ventilation.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Liquid |
Quantity | 250 mL |
Linear Formula | KCl |
Solubility Information | Not miscible in water. |
IUPAC Name | potassium chloride |
Formula Weight | 74.55 |
Concentration or Composition (by Analyte or Components) | 1M aq. soln. |
Chemical Name or Material | Potassium chloride, RNAse free |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'