missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 99+%, for analysis, Thermo Scientific Chemicals

£59.40 - £784.91

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10236350
View Documents Promotion Details
Thermo Scientific Acros
196770010
1 kg Plastic bottle
£59.40
1kg
Please to purchase this item. Need a web account? Register with us today!
10703344
View Documents Promotion Details
Thermo Scientific Acros
196770050
5 kg Plastic bottle
£190.95
5kg
Please to purchase this item. Need a web account? Register with us today!
15169223
View Documents
Thermo Scientific Acros
196770250
25 kg Fibre drum
£784.91
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
Specifications

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
[Cl-].[K+]
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588

Specifications

1420.0°C
7447-40-7
Liquid
ClK
MFCD00011360
15,7742
Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
[Cl-].[K+]
74.55
CHEBI:32588
≥99%
0.5 % max. (1 g, 105°C, 3 hrs)
770.0°C
Colorless
99.0 to 100.5% (on dried substance) (Argentometry)
KCl
06,48
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4873
74.54
Analytical
Potassium chloride, p.a.
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

missing translation for 'einecsNumber' : 231-211-8

missing translation for 'rtecsNumber' : TS8050000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only