missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Potassium chloride, 99%, Molecular Biology Grade Product Code.: 15496249

Thermo Scientific Chemicals Potassium chloride, 99%, Molecular Biology Grade

Product Code. 15496249
500 g
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15496249

Brand: Thermo Scientific Chemicals J64189.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1500°C (sublimation)
Melting Point 773°C
Color White
Physical Form Powder
Quantity 500 g
Merck Index 14,7621
IUPAC Name potassium;chloride
Formula Weight 74.55
Percent Purity 99%
Grade Molecular Biology
Odor Odorless
Sensitivity Hygroscopic
Density 1.984 g/mL
Chemical Name or Material Potassium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ17K001 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberJ64189.A1,J64189.36
Lot NumberU02K020 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberJ64189.A1,J64189.36
Lot NumberP28K020 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ64189.A1,J64189.36
Lot NumberZ11J031 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog NumberJ64189.A1,J64189.36
Lot NumberZ19I032 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog NumberJ64189.A1,J64189.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Potassium chloride, 99%, Molecular Biology Grade >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.