missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, ACS Reagent, Honeywell Fluka™

Product Code. 15688020
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
4 x 2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, 99.0-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Quantity 2.5 kg
Linear Formula KCl
UN Number NONH for all modes of transport
Formula Weight 74.55g/mol
Percent Purity 99 to 100.5%
Grade ACS Reagent
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium chloride
Hazard Statement
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'