missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Crystal, U.S.P. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
Physical Form | Crystals |
Quantity | 2.5 kg |
IUPAC Name | potassium chloride |
Formula Weight | 74.55 |
Grade | FCC/USP |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.99kg |
Chemical Name or Material | Potassium Chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'