missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Crystal, Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- For Molecular Biology Applications
Specifications
Specifications
Physical Form | Crystals |
Quantity | 500 g |
IUPAC Name | potassium;chloride |
Formula Weight | 74.55 |
Grade | Ultrapure Bioreagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.99kg |
Chemical Name or Material | Potassium Chloride |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction