Learn More
Potassium chloride, Optical Grade
Description
Potassium chloride is a salt commonly used in the preparation of phosphate buffered saline and for the extraction and solubilization of proteins. It is a good potassium source. Also used in the hypotonic KCl-propidium iodide protocol for univariate analysis of metaphase chromosomes ; dissociation of eukaryotic ribosomes into subunits by high concentrations of potassium (0.3-1.0 M); preparation of mitochondria, provides an ionic osmotic support for those tissues that assume a gelatinous consistency upon homogenization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1500°C (sublimation) |
| Melting Point | 773°C |
| Physical Form | Broken Crystals |
| Quantity | 100 g |
| Merck Index | 14,7621 |
| Solubility Information | Soluble in water (340mg/ml at 20°C),alcohol and ethanol (1g/250 mL). Insoluble in ether,and acetone. |
| Formula Weight | 74.55 |
| Grade | Optical |
| Odor | Odorless |
| Refractive Index | 1.49 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.