missing translation for 'onlineSavingsMsg'
Learn More
Potassium Chloride, puriss. p.a., Honeywell Fluka™ Product Code.: 15643320

Potassium Chloride, puriss. p.a., Honeywell Fluka™

Product Code. 15643320 Shop All Honeywell Fluka Products
1 kg
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15643320

Brand: Honeywell-Fluka 312481KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.5-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Quantity 1 kg
Linear Formula KCl
UN Number NONH for all modes of transport
IUPAC Name potassium chloride
Formula Weight 74.55g/mol
Percent Purity 99.5 to 100.5%
Grade puriss. p.a.
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium Chloride, Reag. Ph. Eur. reag. ISO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium Chloride, puriss. p.a., Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.