Learn More
Potassium Chloride Solution 3M, Pure, Fisher Chemical™
ClK, CAS Number-7447-40-7, slow-k, potassium chloride, muriate of potash, klor-con, enseal, sylvite, klotrix, potassium chloride kcl, kalitabs, chlorvescent, 1L, CHEBI:32588, Undesignated, WCUXLLCKKVVCTQ-UHFFFAOYSA-M, potassium;chloride, 11360, 74.548, Plastic container, Liquid, 4873, [Cl-].[K+]
Brand: Fisher Chemical J/2892/15
73.62 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 21
Additional Details : CAS Number : 7447-40-7 Weight : 1.25000kg
Description
ClK, CAS Number-7447-40-7, slow-k, potassium chloride, muriate of potash, klor-con, enseal, sylvite, klotrix, potassium chloride kcl, kalitabs, chlorvescent, 1L, CHEBI:32588, Undesignated, WCUXLLCKKVVCTQ-UHFFFAOYSA-M, potassium;chloride, 11360, 74.548, Plastic container, Liquid, 4873, [Cl-].[K+]
Specifications
7447-40-7 | |
1 L | |
MFCD00011360 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
Pure |
Liquid | |
ClK | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
Plastic container |