missing translation for 'onlineSavingsMsg'
Learn More
Potassium chloride, Spectroscopy Grade, Thermo Scientific Chemicals Product Code.: 15491557

Potassium chloride, Spectroscopy Grade, Thermo Scientific Chemicals

Product Code. 15491557
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 15491557

Brand: Thermo Scientific Alfa Aesar 039795.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

 

It is used to replenish electrolytes and restore water electrolyte balance in treating hypokalemia. It is also used in buffers, fertilizers and explosives and finds useful advantages in medicine, scientific applications, and food processing. It plays an important role for the extraction and solubilization of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1500°C (sublimation)
Melting Point 773°C
CAS 7447-40-7
Color White
Physical Form Powder
Quantity 100 g
Molecular Formula ClK
MDL Number MFCD00011360
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
Solubility Information Soluble in water,glycerol,alkalis. Insoluble in ether.
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.55
Grade Spectroscopy
Odor Odorless
Refractive Index 1.49
Appearance White Powder
Density 1.984 g/cm3 at 20°C
Chemical Name or Material Potassium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ30H870 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number039795.22
Lot NumberS02C865 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039795.22
Lot NumberX03C803 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039795.22
Lot NumberT24G823 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039795.22
Lot NumberL14Y029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039795.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium chloride, Spectroscopy Grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.