missing translation for 'onlineSavingsMsg'
Learn More

Potassium chromate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5%

£58.81 - £2794.39

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15613330
View Documents Promotion Details
Honeywell-Fluka
31250-250G
250 g
£58.81
250g
Please to purchase this item. Need a web account? Register with us today!
15603330
View Documents Promotion Details
Honeywell-Fluka
31250-1KG
1 kg
£197.02
1kg
Please to purchase this item. Need a web account? Register with us today!
15633330
View Documents Promotion Details
Honeywell-Fluka
31250-6X1KG
6 x 1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15623330
View Documents Promotion Details
Honeywell-Fluka
31250-50KG
50 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7789-00-6
194.19
XMXNVYPJWBTAHN-UHFFFAOYSA-N
24597
[K+].[K+].[O-][Cr]([O-])(=O)=O
CrK2O4
MFCD00011368
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
CHEBI:75249

Specifications

7789-00-6
K2CrO4
3288
XMXNVYPJWBTAHN-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)chromium
24597
194.19g/mol
ACS/puriss. p.a.
Potassium Chromate, Reag. Ph. Eur.
CrK2O4
MFCD00011368
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
[K+].[K+].[O-][Cr]([O-])(=O)=O
194.19
CHEBI:75249
≥99.5%
Vapor Density: 6.7 (vs air)
Videos
Safety and Handling

Safety and Handling

P201-P273-P280-P301 + P310 + P330-P304 + P340 + P312-P308 + P313

H301-H315-H317-H319-H335-H340-H350i-H410

missing translation for 'einecsNumber' : 232-140-5

missing translation for 'rtecsNumber' : GB2940000

Product Identifier
  • Potassium chromate

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350i-May cause cancer by inhalation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : potassium chromate

SDS
Documents

Documents

Product Certifications