Learn More
Potassium Chromate solution EU Pharmacopoeia, Fisher Chemical™
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Brand: Fisher Chemical RP 160 1000
Code : 04
Additional Details : Weight : 1.00000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Specifications
Bottle | |
EU Ph Ref: 1069201 | |
Potassium Chromate |
1000 mL | |
Eu. Ph. |
Safety and Handling
- Potassium chromate solution. European Pharmacopoeia Ref: 1069201 (RP 160)
Signal Word
- Danger
Hazard Category
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .