missing translation for 'onlineSavingsMsg'
Learn More
Potassium, chunks, in mineral oil, 98%, Thermo Scientific Chemicals Product Code.: 15498979

Potassium, chunks, in mineral oil, 98%, Thermo Scientific Chemicals

Product Code. 15498979
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15498979

Brand: Thermo Scientific Alfa Aesar L13267.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium forms a blue solution in tetrahydrofoliate which is used for the ? -monoalkylation of -lactones in the presence of 18-crown-6. It is used for generation of potassium ethoxide in solution, for use in the Birch reduction of an aromatic ring, for preparation of potassium amide,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium forms a blue solution in tetrahydrofoliate which is used for the ɑ -monoalkylation of -lactones in the presence of 18-crown-6. It is used for generation of potassium ethoxide in solution, for use in the Birch reduction of an aromatic ring, for preparation of potassium amide,

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-09-7
Molecular Formula K
Molecular Weight (g/mol) 39.10
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
Synonym kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atomShow MoreShow Less
PubChem CID 5462222
ChEBI CHEBI:26216
IUPAC Name potassium
SMILES [K]

Specifications

Color Gray-Silver
Melting Point 63°C to 64°C
Boiling Point 765°C to 770°C
Density 0.860 g/mL
UN Number UN2257
Quantity 25 g
Merck Index 14,7604
Solubility Information Reacts with water.
Formula Weight 39.1
Percent Purity 98%
Odor Odorless
Sensitivity Air and moisture sensitive
Form Solid
Chemical Name or Material Potassium, chunks, in mineral oil
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249788 Certificate TypeCertificate of Analysis Date23/02/2024 Catalog NumberL13267.14,L13267.22
Lot Number10240969 Certificate TypeCertificate of Analysis Date31/05/2022 Catalog NumberL13267.14,L13267.22
Lot Number10209714 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13267.14,L13267.22
Lot Number10206909 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL13267.14,L13267.22
Lot Number10209713 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13267.14,L13267.22
compliance-icons compliance-icons
Product Identifier
  • Potassium, chunks, in mineral oil
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Potassium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium, chunks, in mineral oil, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.