missing translation for 'onlineSavingsMsg'
Learn More

Potassium citrate tribasic monohydrate, 99%, extra pure

Product Code. p-4380105 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 12045350

Brand: Thermo Scientific Acros 244210010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6100-05-6 | C6H5K3O7·H2O | 324.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H5K3O7·H2O
Molecular Weight (g/mol) 324.42
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PubChem CID 2735208
ChEBI CHEBI:64746
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]

Specifications

Melting Point 180.0°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula KO2CCH2C(OH)(CO2K)CH2CO2K·H2O
Quantity 1 kg
Merck Index 15,7744
Solubility Information Solubility in water: 640g/L (25°C).
Formula Weight 324.42
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Potassium citrate tribasic monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.