missing translation for 'onlineSavingsMsg'
Learn More
Potassium cyanide, 97%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10143392

Potassium cyanide, 97%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10143392
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10143392

Brand: Thermo Scientific Acros 316450250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium frenchShow MoreShow Less
PubChem CID 9032

Specifications

Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 97%
Linear Formula CKN
Fieser 05,553; 07,299; 08,409; 10,324; 11,433; 13,87; 16,22
Merck Index 15, 7747
SMILES [C-]#N.[K+]
IUPAC Name potassium;cyanide
Molecular Weight (g/mol) 65.116
Formula Weight 65.12
Percent Purity ≥96.0%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Potassium cyanide, 96+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0385556 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number316451000,316450025,316455000,316450250
Lot NumberA0464142 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number316451000,316450025,316450250,316455000
Lot NumberA0432703 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog Number316451000,316450025,316450250,316455000
Lot NumberA0456981 Certificate TypeCertificate of Analysis Date16/10/2023 Catalog Number316451000,316450025,316450250,316455000
Lot NumberA0444516 Certificate TypeCertificate of Analysis Date26/07/2022 Catalog Number316451000,316450025,316450250,316455000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium cyanide, 97%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.