missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, 98+%, for analysis, Thermo Scientific Chemicals

Product Code. 10247403 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
250g
500g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10247403

Brand: Thermo Scientific Acros 196600025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.12
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym cyanide of potassium, zyankali, potassium cyanide, caswell no. 688a, unii-mqd255m2zo, potassium cyanide k cn, kalium cyanid german, cyanure de potassium french, rcra waste number p098
PubChem CID 9032

Specifications

Boiling Point 1625.0°C
Melting Point 634.0°C
Color Colorless
Physical Form Powder/Solid
Quantity 2.5 kg
Assay Percent Range 97% min. (Argentometry)
Linear Formula CKN
Fieser 05,553; 07,299; 08,409; 10,324; 11,433; 13,87; 16,22
Merck Index 15, 7747
Solubility Information Solubility aq. soln.: 400 g/L (20°C). Other solubilities: ethanol 4.5 g/L (19.5°C)
SMILES [K+].[C-]#N
IUPAC Name potassium iminomethanide
Formula Weight 65.12
Percent Purity ≥97%
Grade 97+%
Packaging Plastic bottle
Chemical Name or Material Potassium cyanide, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.