missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent

Specifications
Specifications
Boiling Point | 1625.0°C |
Melting Point | 634.0°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 2.5 kg |
Assay Percent Range | 97% min. (Argentometry) |
Linear Formula | CKN |
Merck Index | 15,7747 |
Solubility Information | Solubility aq. soln.: 400 g/L (20°C). Other solubilities: ethanol 4.5 g/L (19.5°C) |
SMILES | [K+].[C-]#N |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466618 | Certificate of Analysis | 09/10/2024 | 388311000,388310100,388310025,388315000 |
A0466619 | Certificate of Analysis | 09/10/2024 | 388311000,388310100,388310025,388315000 |
A0466620 | Certificate of Analysis | 09/10/2024 | 388311000,388310100,388310025,388315000 |
A0462142 | Certificate of Analysis | 23/04/2024 | 388311000,388310100,388310025,388315000 |
A0458859 | Certificate of Analysis | 20/12/2023 | 388311000,388310100,388310025,388315000 |
Safety and Handling




Product Identifier
- Potassium cyanide
- Danger
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH032-Contact with acids liberates very toxic gas.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction