Learn More
Potassium cyanide, ACS, 96.0% min, Thermo Scientific Chemicals
Description
KCN is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von Richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1625°C |
Melting Point | 634°C |
Physical Form | Granules |
Quantity | 50 g |
Assay Percent Range | 96.0% min. |
Beilstein | 4652394 |
Solubility Information | Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol. |
Formula Weight | 65.12 |
Grade | ACS Reagent |
Sensitivity | Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
62500081 | Certificate of Analysis | 13/02/2025 | 012136.A1,012136.30,012136.18 |
M30L012 | Certificate of Analysis | 03/02/2025 | 012136.A1,012136.30,012136.18 |
S24K028 | Certificate of Analysis | 30/07/2024 | 012136.A1,012136.30,012136.18 |
M30G013 | Certificate of Analysis | 22/07/2022 | 012136.A1,012136.30,012137.36,012136.18 |
K18T025 | Certificate of Analysis | 17/11/2021 | 012136.A1,012136.30,012137.36,012136.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.