missing translation for 'onlineSavingsMsg'
Learn More

Potassium dichromate, 99.5%, for analysis, Thermo Scientific Chemicals

£16.68 - £272.36

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.19
MDL Number MFCD00011367
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10092560
View Documents Promotion Details
Thermo Scientific Acros
196590500
50 g Plastic bottle
£16.68
50g
Please to purchase this item. Need a web account? Register with us today!
10563751
View Documents
Thermo Scientific Acros
196595000
500 g Plastic bottle
£96.18
500g
Please to purchase this item. Need a web account? Register with us today!
10626972
View Documents
Thermo Scientific Acros
196590020
2 kg Plastic bottle
£272.36
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
Specifications

Chemical Identifiers

7778-50-9
294.19
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Cr2K2O7
MFCD00011367
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
CHEBI:53444
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

500.0°C
7778-50-9
Fine Crystals or Crystalline Powder
Cr2K2O7
MFCD00011367
15, 7748
Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
294.19
CHEBI:53444
99.5%
0.05% max. (105°C)
398.0°C
Orange
99.5% min. (Iodometry)
K2Cr2O7
08,410; 09,383; 10,324; 12,405
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24502
294.19
Analytical
Potassium dichromate, p.a.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficu

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-906-6

missing translation for 'rtecsNumber' : HX7680000

missing translation for 'tsca' : TSCA

Product Identifier
  • Potassium dichromate

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only