missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium dichromate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8%
£63.02 - £2850.28
Chemical Identifiers
CAS | 7778-50-9 |
---|---|
Molecular Formula | Cr2K2O7 |
Molecular Weight (g/mol) | 294.182 |
InChI Key | KMUONIBRACKNSN-UHFFFAOYSA-N |
Synonym | potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash |
PubChem CID | 24502 |
ChEBI | CHEBI:53444 |
IUPAC Name | dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
SMILES | [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15673340
|
Honeywell-Fluka
31255-250G |
250 g |
£63.02
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15663340
|
Honeywell-Fluka
31255-1KG |
1 kg |
£184.51
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15683340
|
Honeywell-Fluka
31255-50KG |
50 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7778-50-9 | |
294.182 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
CHEBI:53444 | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] |
Cr2K2O7 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
24502 | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Specifications
7778-50-9 | |
K2Cr2O7 | |
3086 | |
KMUONIBRACKNSN-UHFFFAOYSA-N | |
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24502 | |
294.18g/mol | |
ACS/puriss. p.a. |
Cr2K2O7 | |
MFCD00011367 | |
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash | |
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+] | |
294.182 | |
CHEBI:53444 | |
≥99.8% | |
Potassium Dichromate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P201-P280-P304 + P340 + P310-P305 + P351 + P338-P308 + P313
H272-H301-H312-H314-H317-H330-H334-H340-H350-H360FD-H372-H410
missing translation for 'einecsNumber' : 231-906-6
missing translation for 'rtecsNumber' : HX7680000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium dichromate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Oxidising solid Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H330-Fatal if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : potassium dichromate