missing translation for 'onlineSavingsMsg'
Learn More

Potassium dihydrogen citrate hydrate, 99% (dry basis), Thermo Scientific Chemicals

£62.71 - £175.88

Chemical Identifiers

CAS 866-83-1
Molecular Formula C6H7KO7
Molecular Weight (g/mol) 230.21
MDL Number MFCD00036472
InChI Key WKZJASQVARUVAW-UHFFFAOYNA-M
Synonym potassium dihydrogen citrate
PubChem CID 73896894
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;potassium
SMILES [K+].OC(=O)CC(O)(CC([O-])=O)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11478610
View Documents
Thermo Scientific Alfa Aesar
044415.36
500 g
£62.71
500g
Please to purchase this item. Need a web account? Register with us today!
11488610
View Documents
Thermo Scientific Alfa Aesar
044415.A3
2 kg
£175.88
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium dihydrogen citrate hydrate is used as a buffering agent and a chelating agent. It is used as an alkalizing agent in the renal function. It is found that it increases the bone density, thereby used in research experiments related to the treatment of osteoporosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

866-83-1
230.21
WKZJASQVARUVAW-UHFFFAOYNA-M
73896894
[K+].OC(=O)CC(O)(CC([O-])=O)C(O)=O
C6H7KO7
MFCD00036472
potassium dihydrogen citrate
2-hydroxypropane-1,2,3-tricarboxylic acid;potassium

Specifications

866-83-1
C6H7KO7
4772973
potassium dihydrogen citrate
WKZJASQVARUVAW-UHFFFAOYNA-M
2-hydroxypropane-1,2,3-tricarboxylic acid;potassium
73896894
99%
(dry basis)
Chunks and/or Powder
MFCD00036472
14,7624
Soluble in water. Slightly soluble in ethanol
[K+].OC(=O)CC(O)(CC([O-])=O)C(O)=O
230.21
230.22 (Anhydrous)
Hygroscopic
Potassium dihydrogen citrate hydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 212-753-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.