missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Dihydrogen Orthophosphate, Extra Pure, SLR, Fisher Chemical™
£78.90 - £392.00
Chemical Identifiers
CAS | 7778-77-0 |
---|---|
Molecular Formula | H2KO4P |
Molecular Weight (g/mol) | 136.08 |
MDL Number | MFCD00011401 MFCD00147253 |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
PubChem CID | 516951 |
ChEBI | CHEBI:63036 |
SMILES | [K+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10469180
|
Fisher Chemical
P/4760/53 |
500 g |
£78.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10479180
|
Fisher Chemical
P/4760/60 |
1 kg |
£136.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10756491
|
Fisher Chemical
P/4760/63 |
3 kg |
£364.00
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403753
|
Fisher Chemical
P/4760/65 |
5 kg |
£392.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium Dihydrogen Orthophosphate, Extra Pure, SLR
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
[K+].OP(O)([O-])=O |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
CHEBI:63036 |
Specifications
253°C | |
White | |
Solid | |
MFCD00011401 MFCD00147253 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium;dihydrogen phosphate | |
516951 | |
136.09g/mol |
7778-77-0 | |
4.5 | |
H2KO4P | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 | |
Plastic powder jar |