missing translation for 'onlineSavingsMsg'
Learn More
Potassium dihydrogen phosphate, ACS, 99.0% min, Thermo Scientific Chemicals Product Code.: 11307337

Potassium dihydrogen phosphate, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11307337
500 g
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 11307337

Brand: Thermo Scientific Alfa Aesar 011594.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Buffers For pH Determination

In buffers for pH determination.Potassium dihydrogen phosphate is used as a food additive, fertilizer (NPK), paint additive, coating additive, adhesive, buffering agent and in optical modulators. It is extensively used in research and analytical laboratories as a buffer in mobile/sample preparations for analysis by high pressure liquid chromatography (HPLC) and also in organic synthesis. MKP, monopotassium phosphate, owing to its non-linear optical properties, is used in optical modulators and for second-harmonic generation (SHG).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253°C
Color White
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Slightly soluble in water. Insoluble in alcohol
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09
Grade ACS
Sensitivity Hygroscopic
Appearance White crystals
Density 2.338
Chemical Name or Material Potassium dihydrogen phosphate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium dihydrogen phosphate, ACS, 99.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.