missing translation for 'onlineSavingsMsg'
Learn More

Potassium diphosphate, 98%, extra pure, Thermo Scientific Chemicals

£16.08 - £88.34

Chemical Identifiers

CAS 7320-34-5
Molecular Formula K4O7P2
Molecular Weight (g/mol) 330.33
MDL Number MFCD00011393
InChI Key RYCLIXPGLDDLTM-UHFFFAOYSA-J
Synonym potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphateShow MoreShow Less
PubChem CID 23740
IUPAC Name tetrapotassium;phosphonato phosphate
SMILES [K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10378860
SDS View Documents
Thermo Scientific Acros
269230250
25 g Glass Bottle
£16.08
25g
Please to purchase this item. Need a web account? Register with us today!
10411101
SDS View Documents
Thermo Scientific Acros
269230010
1 kg Plastic bottle
£44.42
1kg
Please to purchase this item. Need a web account? Register with us today!
10454195
SDS View Documents
Thermo Scientific Acros
269230025
2.5 kg Plastic bottle
£88.34
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 230-785-7

Product Identifier
  • Potassium diphosphate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium diphosphate, 98%, extra pure, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.