missing translation for 'onlineSavingsMsg'
Learn More

Potassium disulfitopalladate(II), Premion™, 99.95% (metals basis), Pd 30.4% min, Thermo Scientific™

£127.00 - £479.00

Chemical Identifiers

CAS 68310-13-4
Molecular Formula KO3PdS
Molecular Weight (g/mol) 225.58
MDL Number MFCD00792331
InChI Key FULLCBDUCZPCNB-UHFFFAOYSA-L
Synonym Potassium palladium sulfite
IUPAC Name potassium palladium sulfite
SMILES [K+].[Pd].[O-]S([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389407
View Documents
Thermo Scientific Alfa Aesar
041011.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11399407
View Documents
Thermo Scientific Alfa Aesar
041011.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

68310-13-4
225.58
FULLCBDUCZPCNB-UHFFFAOYSA-L
potassium palladium sulfite
KO3PdS
MFCD00792331
Potassium palladium sulfite
[K+].[Pd].[O-]S([O-])=O

Specifications

68310-13-4
KO3PdS
Potassium palladium sulfite
FULLCBDUCZPCNB-UHFFFAOYSA-L
potassium palladium sulfite
344.75
Premion™
(metals basis)
Powder
MFCD00792331
Soluble in water.
[K+].[Pd].[O-]S([O-])=O
225.58
99.95%
Pd 30.4% min.
Potassium disulfitopalladate(II)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 269-702-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications