missing translation for 'onlineSavingsMsg'
Learn More

Potassium ferricyanide, 99+%, ACS reagent, Thermo Scientific Chemicals

£19.80 - £197.99

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10556412
View Documents Promotion Details
Thermo Scientific Acros
424120050
5 g Glass Bottle
£19.80
5g
Please to purchase this item. Need a web account? Register with us today!
10117710
View Documents Promotion Details
Thermo Scientific Acros
424125000
500 g Plastic bottle
£66.03
500g
Please to purchase this item. Need a web account? Register with us today!
11944321
View Documents
Thermo Scientific Acros
424120025
2.5 kg Plastic bottle
£197.99
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

13746-66-2
329.25
QMTKJUMXUDIUAQ-UHFFFAOYSA-N
26250
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
C6FeK3N6
MFCD00011392
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
tripotassium hexacyanoirontris(ylium)

Specifications

13746-66-2
Crystals and/or Chunks
C6FeK3N6
MFCD00011392
15,7751
Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
329.25
329.26
ACS Reagent
Potassium ferricyanide, 99+%
Orange to Red
99% min. (Iodometry)
K3Fe(CN)6
01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
QMTKJUMXUDIUAQ-UHFFFAOYSA-N
tripotassium hexacyanoirontris(ylium)
26250
≥99%
1.85
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.

GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

missing translation for 'einecsNumber' : 237-323-3

missing translation for 'rtecsNumber' : LJ8225000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.