Learn More
Potassium ferricyanide, 99+%, for analysis, Thermo Scientific Chemicals
£22.90 - £199.00
Chemical Identifiers
CAS | 13746-66-2 |
---|---|
Molecular Formula | C6FeK3N6 |
Molecular Weight (g/mol) | 329.25 |
MDL Number | MFCD00011392 |
InChI Key | QMTKJUMXUDIUAQ-UHFFFAOYSA-N |
Synonym | potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559 |
PubChem CID | 26250 |
SMILES | [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10414583
|
Thermo Scientific Acros
223111000 |
100 g | Glass bottle |
£22.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266370
|
Thermo Scientific Acros
223115000 |
500 g | Glass bottle |
£66.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10390734
|
Thermo Scientific Acros
223110025 |
2.5 kg | Plastic bottle |
£199.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
13746-66-2 | |
329.25 | |
QMTKJUMXUDIUAQ-UHFFFAOYSA-N | |
26250 |
C6FeK3N6 | |
MFCD00011392 | |
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559 | |
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N |
Specifications
13746-66-2 | |
Crystals and/or Chunks | |
C6FeK3N6 | |
MFCD00011392 | |
15, 7751 | |
Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol | |
[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N | |
329.25 | |
329.26 | |
Analytical | |
Potassium ferricyanide, p.a. |
Orange to Red | |
99% min. (Iodometry) | |
K3Fe(CN)6 | |
01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255 | |
potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559 | |
QMTKJUMXUDIUAQ-UHFFFAOYSA-N | |
tripotassium;iron(3+);hexacyanide | |
26250 | |
≥99% | |
1.85 |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Contact with acids liberates very toxic gas.
GHS P Statement
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
missing translation for 'einecsNumber' : 237-323-3
missing translation for 'rtecsNumber' : LJ8225000
missing translation for 'tsca' : TSCA
RUO – Research Use Only