missing translation for 'onlineSavingsMsg'
Learn More
Potassium ferrocyanide trihydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10216460

Potassium ferrocyanide trihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10216460
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10216460

Brand: Thermo Scientific Acros 211095000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Potassium hexacyanoferrate(II)
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 70.0°C
Color Yellow
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula K4Fe(CN)6·3H2O
Merck Index 15, 7752
Solubility Information Solubility in water: 270g/L (12°C). Other solubilities: insoluble in alcohol
Formula Weight 422.4
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 1.85
Density 1.8500g/mL
Chemical Name or Material Potassium ferrocyanide trihydrate, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465594 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number211090025,211095000
Lot NumberA0461139 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number211090025,211095000
Lot NumberA0457624 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog Number211090025,211095000
Lot NumberA0452405 Certificate TypeCertificate of Analysis Date24/04/2023 Catalog Number211090025,211095000
Lot NumberA0421295 Certificate TypeCertificate of Analysis Date02/02/2023 Catalog Number211090025,211095000
Product Identifier
  • Potassium ferrocyanide trihydrate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Potassium ferrocyanide trihydrate, Ferrate(4-), hexakis(cyano-C)-, tetrapotassium, (OC-6-11)-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium ferrocyanide trihydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.