missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10482201
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
SMILES [F-].[K+]

Specifications

Boiling Point 1505°C
Melting Point 860°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 25 g
Assay Percent Range 99%
Linear Formula KF
Fieser 01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282
Merck Index 15, 7753
Solubility Information Solubility in water: 920g/L (18°C). Other solubilities: soluble in hydrofluoric acid,insoluble in alcohol unless water is present
IUPAC Name potassium;fluoride
Formula Weight 58.09
Percent Purity 99%
Grade Extra Pure
Identification Pass Test
Packaging Plastic bottle
Density 2.48
Chemical Name or Material Potassium fluoride, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.