missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, anhydrous, 99%, Thermo Scientific Chemicals

£30.15 - £188.94

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
SMILES [F-].[K+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317737
View Documents
Thermo Scientific Alfa Aesar
014130.18
50 g
£30.15
50g
Please to purchase this item. Need a web account? Register with us today!
11327737
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014130.30
250 g
£69.14
250g
Please to purchase this item. Need a web account? Register with us today!
11307737
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014130.A1
1 kg
£188.94
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium fluoride is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
[F-].[K+]
FK
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
CHEBI:66872

Specifications

1505°C
7789-23-3
Crystalline
FK
3902818
Soluble in water and aqueous hydrogen fluoride. Insoluble in alcohol.
[F-].[K+]
58.10
CHEBI:66872
Odorless
Hygroscopic
0.13 hPa at 752°C
Potassium fluoride, anhydrous
859°C
White
99%
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
522689
58.1
1.363
White various forms (powder/flake/crystalline/beads, etc.)
2.48 g/cm3
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P361-P363-P501c

H301+H311+H331-H318

missing translation for 'dotInformation' : Hazard Class: 6.1, Packing Group: III

missing translation for 'einecsNumber' : 232-151-5

missing translation for 'rtecsNumber' : TT0700000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Potassium fluoride, anhydrous

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Potassium fluoride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only