missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium fluoride, Thermo Scientific Chemicals
£257.47
Chemical Identifiers
CAS | 7789-23-3 |
---|---|
Molecular Formula | FK |
Molecular Weight (g/mol) | 58.10 |
MDL Number | MFCD00011398 |
InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
Synonym | potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 |
PubChem CID | 522689 |
ChEBI | CHEBI:66872 |
SMILES | [F-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15289686
|
Thermo Scientific Acros
449650025 |
2.5 kg | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-23-3 | |
58.10 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
522689 | |
[F-].[K+] |
FK | |
MFCD00011398 | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
CHEBI:66872 |
Specifications
7789-23-3 | |
MFCD00011398 | |
NROKBHXJSPEDAR-UHFFFAOYSA-M | |
potassium fluoride | |
522689 | |
58.09 | |
Potassium fluoride |
FK | |
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
[F-].[K+] | |
58.10 | |
CHEBI:66872 | |
ACS Reagent |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title