missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium fluorosulfate, 99.5%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Description
Use as primary and secondary intermediate. Finds application in chemical research..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 350°C (decomposition) |
Color | White |
Physical Form | Lump/Powder |
Quantity | 50 g |
Linear Formula | KSO3F |
UN Number | UN3260 |
Solubility Information | Hydrolysis. |
Formula Weight | 138.16 |
Percent Purity | 99.5% |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'