Learn More
Potassium fluorosulfate, 99.5%, Thermo Scientific Chemicals
£122.00 - £561.00
Chemical Identifiers
CAS | 13455-22-6 |
---|---|
Molecular Formula | FKO3S |
Molecular Weight (g/mol) | 138.15 |
MDL Number | MFCD00216618 |
InChI Key | VXJAYNWISQFORV-UHFFFAOYSA-M |
Synonym | potassium fluorosulfate, potassium sulfurofluoridate, potassium fluorosulphate, fluorosulfonyloxypotassium, acmc-1c0t1, potassium fluorosulfonyl oxidanide, potassium ion o-fluorosulfonyl oxidanidolate, fluorosulfuric acid,potassium salt 8ci,9ci |
PubChem CID | 23678652 |
IUPAC Name | potassium sulfurofluoridate |
SMILES | [K+].[O-]S(F)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363888
|
Thermo Scientific Alfa Aesar
040322.09 |
10 g |
£122.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353888
|
Thermo Scientific Alfa Aesar
040322.18 |
50 g |
£561.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Use as primary and secondary intermediate. Finds application in chemical research..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13455-22-6 | |
138.15 | |
VXJAYNWISQFORV-UHFFFAOYSA-M | |
23678652 | |
[K+].[O-]S(F)(=O)=O |
FKO3S | |
MFCD00216618 | |
potassium fluorosulfate, potassium sulfurofluoridate, potassium fluorosulphate, fluorosulfonyloxypotassium, acmc-1c0t1, potassium fluorosulfonyl oxidanide, potassium ion o-fluorosulfonyl oxidanidolate, fluorosulfuric acid,potassium salt 8ci,9ci | |
potassium sulfurofluoridate |
Specifications
350°C (decomposition) | |
White | |
FKO3S | |
MFCD00216618 | |
potassium fluorosulfate, potassium sulfurofluoridate, potassium fluorosulphate, fluorosulfonyloxypotassium, acmc-1c0t1, potassium fluorosulfonyl oxidanide, potassium ion o-fluorosulfonyl oxidanidolate, fluorosulfuric acid,potassium salt 8ci,9ci | |
VXJAYNWISQFORV-UHFFFAOYSA-M | |
potassium sulfurofluoridate | |
23678652 | |
99.5% | |
Potassium fluorosulfate |
13455-22-6 | |
Lump/Powder | |
KSO3F | |
UN3260 | |
Hydrolysis. | |
[K+].[O-]S(F)(=O)=O | |
138.15 | |
138.16 | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only