Learn More
Potassium formate, 99% (water less than 2%), Thermo Scientific Chemicals
£24.72 - £257.28
Chemical Identifiers
CAS | 590-29-4 |
---|---|
Molecular Formula | CHKO2 |
Molecular Weight (g/mol) | 84.115 |
MDL Number | MFCD00013100 |
InChI Key | WFIZEGIEIOHZCP-UHFFFAOYSA-M |
Synonym | potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent |
PubChem CID | 2735122 |
ChEBI | CHEBI:63316 |
IUPAC Name | potassium;formate |
SMILES | C(=O)[O-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11411938
|
Thermo Scientific Alfa Aesar
A14551.30 |
250 g |
£24.72
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401938
|
Thermo Scientific Alfa Aesar
A14551.0B |
1000 g |
£70.05
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11421938
|
Thermo Scientific Alfa Aesar
A14551.0I |
5000 g |
£257.28
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium formate acts as an intermediate in the formate potash process for the production of potassium. It is a deicing salt for used on roads. It serves as a corrosion inhibitor, an anti-scaling agent, a functional fluid, a processing aid and used in petroleum production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium formate acts as an intermediate in the formate potash process for the production of potassium. It is a deicing salt for used on roads. It serves as a corrosion inhibitor, an anti-scaling agent, a functional fluid, a processing aid and used in petroleum production.
Solubility
Soluble in water and alcohol. Insoluble in ether.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
590-29-4 | |
84.115 | |
WFIZEGIEIOHZCP-UHFFFAOYSA-M | |
2735122 | |
potassium;formate |
CHKO2 | |
MFCD00013100 | |
potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent | |
CHEBI:63316 | |
C(=O)[O-].[K+] |
Specifications
590-29-4 | |
1.91 | |
CHKO2 | |
3623272 | |
14,7633 | |
Soluble in water and alcohol; Insoluble in ether. | |
C(=O)[O-].[K+] | |
84.115 | |
CHEBI:63316 | |
99% | |
Potassium Formate |
165°C to 168°C | |
99% | |
MFCD00013100 | |
Very Hygroscopic | |
potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent | |
WFIZEGIEIOHZCP-UHFFFAOYSA-M | |
potassium;formate | |
2735122 | |
84.12 | |
(water <2%) |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 209-677-9
missing translation for 'rtecsNumber' : LQ9625000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only