missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachloroosmate(IV), min 39% Os, Thermo Scientific Chemicals

Catalog Number 10348670
98.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
16871-60-6
Cl6K2Os
481.11
MFCD00011371
16871-60-6
Cl6K2Os
481.11
MFCD00011371

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 600.0°C
Color Bordeaux to Purple
Physical Form Powder
Quantity 1 g
Linear Formula K2OsCl6
Merck Index 15, 7756
Solubility Information Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol
Formula Weight 481.11
Percent Purity ≥39% (Os)
Packaging Glass bottle
Density 3.42
Chemical Name or Material Potassium hexachloroosmate(IV), min 39% Os
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hexachloroosmate(IV)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'