Learn More
Potassium hexachloropalladate(IV), Pd 26.3% min, Thermo Scientific Chemicals
£75.20 - £375.00
Chemical Identifiers
CAS | 16919-73-6 |
---|---|
Molecular Formula | Cl6K2Pd |
Molecular Weight (g/mol) | 397.317 |
MDL Number | MFCD00043081 |
InChI Key | OEDRXJBJMMJUSV-UHFFFAOYSA-H |
Synonym | potassium hexachloropalladate iv, dipotassium hexachloropalladate, dipotassium hexachloropalladate 2-, ccris 7827, potassiuim hexachloropalladate iv, palladate 2-, hexachloro-, dipotassium, potassium palladium iv chloride, palladate 2-, hexachloro-, dipotassium, oc-6-11, potassium chloropalladate iv 6ci, dipotassiumhexachloropalladate |
PubChem CID | 61852 |
IUPAC Name | dipotassium;hexachloropalladium(2-) |
SMILES | Cl[Pd-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381968
|
Thermo Scientific Alfa Aesar
011884.03 |
1 g |
£75.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391968
|
Thermo Scientific Alfa Aesar
011884.06 |
5 g |
£375.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium hexachloropalladate(IV) is used as organic reagents and pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16919-73-6 | |
397.317 | |
OEDRXJBJMMJUSV-UHFFFAOYSA-H | |
61852 | |
Cl[Pd-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+] |
Cl6K2Pd | |
MFCD00043081 | |
potassium hexachloropalladate iv, dipotassium hexachloropalladate, dipotassium hexachloropalladate 2-, ccris 7827, potassiuim hexachloropalladate iv, palladate 2-, hexachloro-, dipotassium, potassium palladium iv chloride, palladate 2-, hexachloro-, dipotassium, oc-6-11, potassium chloropalladate iv 6ci, dipotassiumhexachloropalladate | |
dipotassium;hexachloropalladium(2-) |
Specifications
(decomposition) | |
Red | |
Cl6K2Pd | |
UN3077 | |
Soluble in water. | |
Cl[Pd-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+] | |
397.317 | |
397.32 | |
Odorless | |
2.738 g/mL |
16919-73-6 | |
Crystalline | |
MFCD00043081 | |
potassium hexachloropalladate iv, dipotassium hexachloropalladate, dipotassium hexachloropalladate 2-, ccris 7827, potassiuim hexachloropalladate iv, palladate 2-, hexachloro-, dipotassium, potassium palladium iv chloride, palladate 2-, hexachloro-, dipotassium, oc-6-11, potassium chloropalladate iv 6ci, dipotassiumhexachloropalladate | |
OEDRXJBJMMJUSV-UHFFFAOYSA-H | |
dipotassium;hexachloropalladium(2-) | |
61852 | |
Pd 26.3% min. | |
Hygroscopic | |
Potassium hexachloropalladate(IV) |
Safety and Handling
GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 240-974-6
missing translation for 'rtecsNumber' : RT3353000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only